Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenyl phosphate, MilliporeSigma™ Supelco™

Catalog No. 111020624 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0624 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0624 Supplier MilliporeSigma Supelco Supplier No. 442829
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 115-86-6
Vapor Pressure 1.3 mmHg (200°C)
Flash Point 220°C
Physical Form Neat
Linear Formula (C6H5 O)3 PO
MDL Number MFCD00003031
Melting Point 48°C to 50°C
Boiling Point 244°C (10 mmHg)
Solubility Soluble in water
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 326.28
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.