CAS RN 115-90-2
IUPAC Name:
O,O-diethyl O-4-methanesulfinylphenyl phosphorothioate
Synonyms:
fensulfothion VUAgT 108 caswell no. 343 daconit chemagro 25141 VUAgT 96 epa pesticide chemical code 032701 terracur P dasanit
Molecular Weight (g/mol):
308.35
Molecular Formula:
C11H17O4PS2
InChi Key:
XDNBJTQLKCIJBV-UHFFFAOYNA-N
SMILES:
CCOP(=S)(OCC)OC1=CC=C(C=C1)S(C)=O
1
–
1
of
1
results
1
Fensulfothion, SPEX CertiPrep™
CAS: 115-90-2 Molecular Formula: C11H17O4PS2 Molecular Weight (g/mol): 308.347 InChI Key: XDNBJTQLKCIJBV-UHFFFAOYSA-N PubChem CID: 8292 ChEBI: CHEBI:34760 IUPAC Name: diethoxy-(4-methylsulfinylphenoxy)-sulfanylidene-$l^{5}-phosphane SMILES: CCOP(=S)(OCC)OC1=CC=C(C=C1)S(=O)C