accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOH

CAS RN 115016-95-0

IUPAC Name: 2-hydroxy-4-phenylbutanoic acid
Synonyms: 2-hydroxy-4-phenylbutanoic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: JNJCEALGCZSIGB-UHFFFAOYNA-N
SMILES: OC(CCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results