Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-Hydroxy-4-phenylbutyric Acid 98.0+%, TCI America™
SDP

Catalog No. H13395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13395G 5 g
H13391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13395G Supplier TCI America Supplier No. H13395G

Chemical Identifiers

CAS 115016-95-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD06656492
InChI Key JNJCEALGCZSIGB-VIFPVBQESA-N
PubChem CID 11159656
IUPAC Name (2S)-2-hydroxy-4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CCC(C(=O)O)O

Specifications

Melting Point 117°C
Color White
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-2-Hydroxy-4-phenylbutyric Acid
EINECSNumber (3)-4133
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.