accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115066-14-3

IUPAC Name: 7-nitro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile
Synonyms: CNQX
Molecular Weight (g/mol): 232.16
Molecular Formula: C9H4N4O4
InChi Key: RPXVIAFEQBNEAX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=C2NC(=O)C(=O)NC2=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results