CAS RN 1151995-69-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Tubulin Polymerization Inhibitor II
The Tubulin Polymerization Inhibitor II, also referenced under CAS 1151995-69-5, acts as an effective anti-microtubule agent. This small molecule/inhibitor is primarily used for Cell Structure applications.