accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115416-52-9

IUPAC Name: 2-[(5-nitropyridin-2-yl)amino]propan-1-ol
Synonyms: 2-[(5-nitropyridin-2-yl)amino]propan-1-ol
Molecular Weight (g/mol): 197.19
Molecular Formula: C8H11N3O3
InChi Key: IFUIXZIUISLXKT-UHFFFAOYNA-N
SMILES: CC(CO)NC1=NC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results