Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-N-(5-Nitro-2-pyridyl)alaninol 98.0+%, TCI America™
SDP

Catalog No. N0547100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0547100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0547100MG Supplier TCI America Supplier No. N0547100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 115416-52-9
Molecular Formula C8H11N3O3
Molecular Weight (g/mol) 197.19
MDL Number MFCD00145287
InChI Key IFUIXZIUISLXKT-UHFFFAOYNA-N
PubChem CID 44630406
IUPAC Name 2-[(5-nitropyridin-2-yl)amino]propan-1-ol
SMILES CC(CO)NC1=NC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 100°C
Color Yellow
Quantity 100 mg
Formula Weight 197.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-N-(5-Nitro-2-pyridyl)alaninol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.