accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1155-00-6

IUPAC Name: 1-nitro-2-[(2-nitrophenyl)disulfanyl]benzene
Synonyms: disulfide, bis(2-nitrophenyl)
Molecular Weight (g/mol): 308.33
Molecular Formula: C12H8N2O4S2
InChi Key: NXCKJENHTITELM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1SSC1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)