Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-nitrophenyl) disulfide, 98%, Thermo Scientific™

Catalog No. AAB2561522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2561522 100 g
AAB2561514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2561522 Supplier Thermo Scientific Chemicals Supplier No. B2561522

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1155-00-6
Molecular Formula C12H8N2O4S2
Molecular Weight (g/mol) 308.33
MDL Number MFCD00007130
InChI Key NXCKJENHTITELM-UHFFFAOYSA-N
Synonym bis 2-nitrophenyl disulfide, disulfide, bis 2-nitrophenyl, 2,2'-dinitrodiphenyl disulfide, o-nitrophenyl disulfide, 2-nitrophenyl disulfide, o,o'-dinitrodiphenyl disulfide, disulfide, bis o-nitrophenyl, bis o-nitrophenyl disulfide, 1,2-bis 2-nitrophenyl disulfane
PubChem CID 70861
IUPAC Name 1-nitro-2-[(2-nitrophenyl)disulfanyl]benzene
SMILES [O-][N+](=O)C1=CC=CC=C1SSC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 192°C to 196°C
Quantity 100 g
Beilstein 1016770
Formula Weight 308.33
Percent Purity 98%
Chemical Name or Material Bis(2-nitrophenyl) disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.