accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115651-77-9

IUPAC Name: 2-(dibutylamino)-1-phenylpropan-1-ol
Synonyms: 2-(dibutylamino)-1-phenylpropan-1-ol
Molecular Weight (g/mol): 263.43
Molecular Formula: C17H29NO
InChi Key: BRRGNOFUBFINSX-UHFFFAOYNA-N
SMILES: CCCCN(CCCC)C(C)C(O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results