Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2S)-2-(Dibutylamino)-1-phenyl-1-propanol 95.0+%, TCI America™
SDP

Catalog No. D21285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21285G Supplier TCI America Supplier No. D21285G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 115651-77-9
Molecular Formula C17H29NO
Molecular Weight (g/mol) 263.425
MDL Number MFCD00142689
InChI Key BRRGNOFUBFINSX-RDJZCZTQSA-N
Synonym (+)-alpha-[1-(Dibutylamino)ethyl]benzyl Alcohol, (+)-N,N-Dibutylnorephedrin
PubChem CID 9903376
IUPAC Name (1R,2S)-2-(dibutylamino)-1-phenylpropan-1-ol
SMILES CCCCN(CCCC)C(C)C(C1=CC=CC=C1)O

Specifications

Color Yellow
Boiling Point 121°C
Quantity 5 g
Formula Weight 263.43
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (1R,2S)-2-(Dibutylamino)-1-phenyl-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.