accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115822-57-6

OO(R)O

CAS RN 115822-57-6

IUPAC Name: (3R)-3,4-dihydro-2H-1-benzopyran-3-carboxylate
Synonyms: (3R)-3,4-dihydro-2H-1-benzopyran-3-carboxylate
Molecular Weight (g/mol): 177.18
Molecular Formula: C10H9O3
InChi Key: UGAGZMGJJFSKQM-MRVPVSSYSA-M
SMILES: [O-]C(=O)[C@H]1COC2=CC=CC=C2C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results