accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1159-15-5

IUPAC Name: 5-[(diaminomethylidene)amino]-2-(4-methylbenzenesulfonamido)pentanoic acid
Synonyms: 5-[(diaminomethylidene)amino]-2-(4-methylbenzenesulfonamido)pentanoic acid
Molecular Weight (g/mol): 328.39
Molecular Formula: C13H20N4O4S
InChi Key: KFNRNFXZFIRNEO-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(O)=O
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results