Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Tosyl-L-arginine, TCI America™
SDP

Catalog No. T06845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T06845G Supplier TCI America Supplier No. T06845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1159-15-5
Molecular Formula C13H20N4O4S
Molecular Weight (g/mol) 328.39
MDL Number MFCD00019732
InChI Key KFNRNFXZFIRNEO-UHFFFAOYNA-N
Synonym tos-arg-oh, n-tosyl-l-arginine, n-p-tosyl-l-arginine, tosyl-l-arginine, nalpha-tosyl-l-arginine, nalpha-p-toluenesulfonyl-l-arginine, p-tosyl-l-arginine, l-arginine, n2-4-methylphenyl sulfonyl, 2s-5-carbamimidamido-2-4-methylbenzenesulfonamido pentanoic acid, tosyl arginine
PubChem CID 52501
IUPAC Name 5-[(diaminomethylidene)amino]-2-(4-methylbenzenesulfonamido)pentanoic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 328.39
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Tosyl-L-arginine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.