accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115933-30-7

IUPAC Name: 5-(4-fluorophenyl)thiophene-2-carboxylic acid
Synonyms: 5-(4-fluorophenyl)thiophene-2-carboxylic acid
Molecular Weight (g/mol): 222.23
Molecular Formula: C11H7FO2S
InChi Key: YOFXYDVYMHOXSY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(S1)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results