Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Fluorophenyl)thiophene-2-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. F10735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10735G 5 g
F10731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10735G Supplier TCI America Supplier No. F10735G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 115933-30-7
Molecular Formula C11H7FO2S
Molecular Weight (g/mol) 222.233
MDL Number MFCD01316570
InChI Key YOFXYDVYMHOXSY-UHFFFAOYSA-N
Synonym 5-4-fluorophenyl thiophene-2-carboxylic acid, 5-4-fluorophenyl-2-thiophenecarboxylic acid, 2-carboxy-5-4-fluorophenyl thiophene, 2-thiophenecarboxylicacid, 5-4-fluorophenyl, 5-4-fluoro-phenyl-thiophene-2-carboxylicacid, fluorophenylthiophenecarboxylicacid, 5-4-fluorophenyl-2-thenoic acid, 1-5-carboxythien-2-yl-4-fluorobenzene, 5-4-fluorophenyl thiophen-2-carboxylic acid, 5-4-fluorophenyl thiophene-2-carboxylicacid
PubChem CID 1873401
IUPAC Name 5-(4-fluorophenyl)thiophene-2-carboxylic acid
SMILES C1=CC(=CC=C1C2=CC=C(S2)C(=O)O)F

Specifications

Melting Point 250°C
Color White-Yellow
Quantity 5 g
Formula Weight 222.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4-Fluorophenyl)thiophene-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.