accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3CH3H3CCH3(E)(E)(E)(E)

CAS RN 116-31-4

IUPAC Name: (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenal
Synonyms: retinaldehyde
Molecular Weight (g/mol): 284.44
Molecular Formula: C20H28O
InChi Key: NCYCYZXNIZJOKI-OVSJKPMPSA-N
SMILES: C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=O
Grade 
  • (7)
Physical Form 
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (7)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)