Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

all-trans-Retinal, Spectrum™ Chemical
SDP

Catalog No. 18600421 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
100 g
160 g
390 g
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-421 390 g Amber Glass Bottle
18-600-415 100 mg Amber Glass Bottle
18-600-416 250 mg Amber Glass Bottle
18-600-417 1 g Amber Glass Bottle
18-600-418 5 g Amber Glass Bottle
18-600-419 100 g Amber Glass Bottle
18-600-420 160 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-600-421 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. R3041390GMBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

R3041, 116-31-4, C20H28O

Spectrum™ Chemical all-trans-Retinal is also known as retinaldehyde and considered a form of vitamin A. It is the chemical basis of animal vision. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 116-31-4
Molecular Formula C20H28O
Molecular Weight (g/mol) 284.44
InChI Key NCYCYZXNIZJOKI-OVSJKPMPSA-N
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenal
SMILES C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 390 g
Percent Purity 98%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material all-Trans-Retinal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.