accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1160823-76-6

IUPAC Name: octyl 4,6-dibromo-3-fluorothieno[3,4-b]thiophene-2-carboxylate
Synonyms: octyl 4,6-dibromo-3-fluorothieno[3,4-b]thiophene-2-carboxylate
Molecular Weight (g/mol): 472.23
Molecular Formula: C15H17Br2FO2S2
InChi Key: AXLMQOPQPUEOQD-UHFFFAOYSA-N
SMILES: CCCCCCCCOC(=O)C1=C(F)C2=C(Br)SC(Br)=C2S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results