accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3CH3CPH3CH3CCH3CH3H3CCH3OCH3OCH3CH3CH3(?)

CAS RN 1160861-53-9

IUPAC Name: di-tert-butyl[3,6-dimethoxy-2',4',6'-tris(propan-2-yl)-[1,1'-biphenyl]-2-yl]phosphane
Synonyms: di-tert-butyl({2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl})phosphane
Molecular Weight (g/mol): 484.71
Molecular Formula: C31H49O2P
InChi Key: REWLCYPYZCHYSS-UHFFFAOYSA-N
SMILES: COC1=C(P(C(C)(C)C)C(C)(C)C)C(=C(OC)C=C1)C1=C(C=C(C=C1C(C)C)C(C)C)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (1)