accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116195-81-4

NII

CAS RN 116195-81-4

IUPAC Name: 2,5-diiodopyridine
Synonyms: 2,5-diiodopyridine
Molecular Weight (g/mol): 330.9
Molecular Formula: C5H3I2N
InChi Key: YHWFCFIXXMXRBF-UHFFFAOYSA-N
SMILES: IC1=CN=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results