accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOO

CAS RN 1162-65-8

IUPAC Name: 11-methoxy-6,8,19-trioxapentacyclo[10.7.0.0²,⁹.0³,⁷.0¹³,¹⁷]nonadeca-1,4,9,11,13(17)-pentaene-16,18-dione
Synonyms: aflatoxin B1
Molecular Weight (g/mol): 312.28
Molecular Formula: C17H12O6
InChi Key: OQIQSTLJSLGHID-UHFFFAOYNA-N
SMILES: COC1=C2C3=C(C(=O)CC3)C(=O)OC2=C2C3C=COC3OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results