Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aflatoxin B1, crystalline, Thermo Scientific Chemicals

Catalog No. AC227340100
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC227340100 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC227340100 Supplier Thermo Scientific Chemicals Supplier No. 227340100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Aflatoxin B1, crystalline, CAS # 1162-65-8, is a member of a group of mycotoxins presenting potent hepatotoxic and carcinogenic activities.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Aflatoxin B1 (AFB1) is a member of a group of mycotoxins produced by Aspergillus flavus, and Aspergillus parasiticus showing carcinogenic activities
• AFB1 can induce the transversion of G to T in the third position of codon 249 of the p53 tumor suppressor gene, resulting in a mutation and promoting the carcinogenic process

Applications

• Aflatoxin B1 shows teratogenic activity
• It is a CYP1A2, CYP2A6, CYP2D6, and CYP3A substrate and can be used to study the CYP family in in vitro and in vivo studies.
• It can cause immunosuppression in animal models.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1162-65-8
Molecular Formula C17H12O6
Molecular Weight (g/mol) 312.28
MDL Number MFCD00869647
InChI Key OQIQSTLJSLGHID-UHFFFAOYNA-N
Synonym aflatoxin b1, ccris 12, unii-9n2n2y55mh, afb1, 2,3,6aalpha,9aalpha-tetrahydro-4-methoxycyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione, 1h,11h-cyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione,2,3,6a,9a-tetrahydro-4-methoxy-, 6ar,9as, cyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione, 2,3,6a,9a-tetrahydro-4-methoxy, cyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione, 2,3,6a,9a-tetrahydro-4-methoxy-, 6ar,9as, aflatoxin-b1, aflatoxin b
PubChem CID 186907
ChEBI CHEBI:2504
IUPAC Name 11-methoxy-6,8,19-trioxapentacyclo[10.7.0.0²,⁹.0³,⁷.0¹³,¹⁷]nonadeca-1,4,9,11,13(17)-pentaene-16,18-dione
SMILES COC1=C2C3=C(C(=O)CC3)C(=O)OC2=C2C3C=COC3OC2=C1

Specifications

Packaging Glass Bottle
Quantity 10 mg
Merck Index 15, 171
Formula Weight 312.27
Chemical Name or Material Aflatoxin B1
Hazard Category Danger
Hazard Statement GHS H Statement:
Fatal if inhaled.
May cause cancer.
Fatal in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-603-3
RTECSNumber GY1925000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.