accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116241-61-3

IUPAC Name: 2-fluoro-6-methoxypyridine
Synonyms: 2-fluoro-6-methoxypyridine
Molecular Weight (g/mol): 127.12
Molecular Formula: C6H6FNO
InChi Key: PKZOFECOOLMHMG-UHFFFAOYSA-N
SMILES: COC1=NC(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results