accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNHBrHBr

CAS RN 116258-17-4

IUPAC Name: 2-benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide
Synonyms: 2-benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide
Molecular Weight (g/mol): 350.1
Molecular Formula: C12H18Br2N2
InChi Key: SOMPEQIPSQFVMO-UHFFFAOYNA-N
SMILES: Br.Br.C(N1CC2CC1CN2)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results