CAS RN 1164153-22-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ JNK3 Inhibitor XII, SR-3576
The JNK3 Inhibitor XII, SR-3576, also referenced under CAS 1164153-22-3, controls the biological activity of JNK3.