accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNOHOHCl

CAS RN 1164469-60-6

IUPAC Name: 1-[4-(2-methoxyphenyl)piperazin-1-yl]-3-(naphthalen-1-yloxy)propan-2-ol hydrochloride
Synonyms: naftopidil hydrochloride
Molecular Weight (g/mol): 428.96
Molecular Formula: C24H29ClN2O3
InChi Key: VQAAEWMEVIOHTJ-UHFFFAOYNA-N
SMILES: Cl.COC1=CC=CC=C1N1CCN(CC(O)COC2=C3C=CC=CC3=CC=C2)CC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)