Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naftopidil hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. AAJ63249MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63249MF 50 mg
AAJ63249MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63249MF Supplier Thermo Scientific Chemicals Supplier No. J63249MF
Add to Cart

missing translation for 'validMainframeMsg'

An alpha1-adrenoceptor antagonist

An alpha1-adrenoceptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1164469-60-6
Molecular Formula C24H29ClN2O3
Molecular Weight (g/mol) 428.957
MDL Number MFCD09971016
InChI Key VQAAEWMEVIOHTJ-UHFFFAOYSA-N
Synonym naftopidil hydrochloride, opera_id_518, 1-4-2-methoxyphenyl piperazin-1-yl-3-naphthalen-1-yloxypropan-2-ol hydrochloride, 4-2-methoxyphenyl-?-1-naphthalenyloxy methyl-1-piperazineethanol hydrochloride
PubChem CID 6603044
IUPAC Name 1-[4-(2-methoxyphenyl)piperazin-1-yl]-3-naphthalen-1-yloxypropan-2-ol;hydrochloride
SMILES COC1=CC=CC=C1N2CCN(CC2)CC(COC3=CC=CC4=CC=CC=C43)O.Cl

Specifications

Melting Point 116°C to 118°C
Color White
Quantity 50 mg
Solubility Information Soluble in DMSO; Also soluble in ethanol; Insoluble in water.
Formula Weight 428.96
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Naftopidil hydrochloride
RTECSNumber TL9336500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.