accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COH3COH3C

CAS RN 116549-34-9

IUPAC Name: 3,3-dimethoxyhexane
Synonyms: 3,3-dimethoxyhexane
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: KYKSUAPFGPQISC-UHFFFAOYSA-N
SMILES: CCCC(CC)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results