Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethoxyhexane, TCI America™
SDP

Catalog No. D18895ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18895ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D18895ML Supplier TCI America Supplier No. D18895ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 116549-34-9
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00060093
InChI Key KYKSUAPFGPQISC-UHFFFAOYSA-N
Synonym 3-Hexanone Dimethylketal
PubChem CID 568629
IUPAC Name 3,3-dimethoxyhexane
SMILES CCCC(CC)(OC)OC

Specifications

Color Colorless
UN Number 3271
Quantity 5 mL
Formula Weight 146.23
Physical Form Liquid
Chemical Name or Material 3,3-Dimethoxyhexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.