accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNCH3SHNO(E)

CAS RN 1166-34-3

IUPAC Name: hydrogen (2E)-N-(2-{[3-(dimethylamino)propyl]sulfanyl}phenyl)-3-phenylprop-2-enamide chloride
Synonyms: cinanserin
Molecular Weight (g/mol): 376.94
Molecular Formula: C20H25ClN2OS
InChi Key: LXGJPDKYMJJWRB-IERUDJENSA-N
SMILES: [H+].[Cl-].CN(C)CCCSC1=CC=CC=C1NC(=O)\C=C\C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results