Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinanserin hydrochloride, 99+%, Thermo Scientific™

Catalog No. AAJ63370MF
Change view
Click to view available options
Quantity:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63370MF 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63370MF Supplier Thermo Scientific Chemicals Supplier No. J63370MF

missing translation for 'validMainframeMsg'

A 5-HT antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1166-34-3
Molecular Formula C20H25ClN2OS
Quantity 50mg
MDL Number MFCD00055609
Solubility Information Soluble to 50 mM in water
InChI Key LXGJPDKYMJJWRB-IERUDJENSA-N
SMILES CN(C)CCCSC1=CC=CC=C1NC(=O)C=CC2=CC=CC=C2.Cl
IUPAC Name (E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide;hydrochloride
Molecular Weight (g/mol) 376.943
PubChem CID 6433141
Formula Weight 376.94
Percent Purity ≥99%
Chemical Name or Material Cinanserin hydrochloride
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber UC6323000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.