accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1166-52-5

H3COOOHOHOH

CAS RN 1166-52-5

IUPAC Name: dodecyl 3,4,5-trihydroxybenzoate
Synonyms: dodecyl gallate
Molecular Weight (g/mol): 338.44
Molecular Formula: C19H30O5
InChi Key: RPWFJAMTCNSJKK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC(=O)C1=CC(O)=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)