accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116631-94-8

IUPAC Name: 1,4-dimethyl 2,3-dibromobut-2-enedioate
Synonyms: 1,4-dimethyl 2,3-dibromobut-2-enedioate
Molecular Weight (g/mol): 301.92
Molecular Formula: C6H6Br2O4
InChi Key: XMUOGQHVYZDUCF-UHFFFAOYSA-N
SMILES: COC(=O)C(Br)=C(Br)C(=O)OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results