accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHSH3COOOFF

CAS RN 116686-15-8

IUPAC Name: N-[4-acetyl-2-(2,4-difluorophenoxy)phenyl]methanesulfonamide
Synonyms: N-[4-acetyl-2-(2,4-difluorophenoxy)phenyl]methanesulfonamide
Molecular Weight (g/mol): 341.33
Molecular Formula: C15H13F2NO4S
InChi Key: DIIYLGZNZGPXRR-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(NS(C)(=O)=O)C(OC2=CC=C(F)C=C2F)=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results