accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHONHHN(S)H3C

CAS RN 116747-54-7

IUPAC Name: 2-[(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanamido]acetic acid
Synonyms: [(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanamido]acetic acid
Molecular Weight (g/mol): 368.39
Molecular Formula: C20H20N2O5
InChi Key: GOCNEQGFDAXBQE-LBPRGKRZSA-N
SMILES: C[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(=O)NCC(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)