CAS RN 116857-07-9
CAS RN 116857-07-9
IUPAC Name:
(9H-fluoren-9-yl)methyl N-[(3S)-2-oxooxolan-3-yl]carbamate
Synonyms:
9H-fluoren-9-ylmethyl N-[(3S)-2-oxooxolan-3-yl]carbamate
Molecular Weight (g/mol):
323.35
Molecular Formula:
C19H17NO4
InChi Key:
VVPJFGZZHSVGJF-KRWDZBQOSA-N
SMILES:
O=C(N[C@H]1CCOC1=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Homoserine lactone, Advanced ChemTech
CAS: 116857-07-9 Molecular Formula: C19H17NO4 Molecular Weight (g/mol): 323.35 MDL Number: MFCD04112472 InChI Key: VVPJFGZZHSVGJF-KRWDZBQOSA-N IUPAC Name: (9H-fluoren-9-yl)methyl N-[(3S)-2-oxooxolan-3-yl]carbamate SMILES: O=C(N[C@H]1CCOC1=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2