accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNOHOOH

CAS RN 116861-26-8

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxypropanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-hydroxypropanoic acid
Molecular Weight (g/mol): 327.34
Molecular Formula: C18H17NO5
InChi Key: JZTKZVJMSCONAK-UHFFFAOYNA-N
SMILES: OCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results