Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-serine 95.0+%, TCI America™
SDP

Catalog No. F06075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06075G 5 g
F06071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06075G Supplier TCI America Supplier No. F06075G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 116861-26-8
Molecular Formula C18H17NO5
Molecular Weight (g/mol) 327.336
MDL Number MFCD00077068
InChI Key JZTKZVJMSCONAK-MRXNPFEDSA-N
Synonym fmoc-d-ser-oh, fmoc-d-serine, n-9h-fluoren-9-ylmethoxy carbonyl-d-serine, n-fmoc-d-serine, d-serine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-serine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxypropanoic acid, ambotzfaa1508, pubchem10038, n-fmoc-d-ala oh
PubChem CID 688637
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxypropanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CO)C(=O)O

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 5 g
Formula Weight 327.34
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-serine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.