accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBr

CAS RN 116986-13-1

IUPAC Name: 3-(bromomethyl)pyridine-2-carbonitrile
Synonyms: 3-(bromomethyl)pyridine-2-carbonitrile
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: BSSLVCQAYWTRRC-UHFFFAOYSA-N
SMILES: BrCC1=C(N=CC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results