CAS RN 1169942-85-1
CAS RN 1169942-85-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 1-Methyl-1,2,3,6-Tetrahydropyridine-4-Boronic Acid Pinacol Ester
Sigma Aldrich 1-Methyl-1,2,3,6-Tetrahydropyridine-4-Boronic Acid Pinacol Ester
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.