accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOCH3OOCH3

CAS RN 117-47-5

IUPAC Name: 1,3-diethyl 2-(pentan-2-yl)propanedioate
Synonyms: 1,3-diethyl 2-(pentan-2-yl)propanedioate
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: RQFSNEWORATSCC-UHFFFAOYNA-N
SMILES: CCCC(C)C(C(=O)OCC)C(=O)OCC
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results