accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOO

CAS RN 117-79-3

IUPAC Name: 2-amino-9,10-dihydroanthracene-9,10-dione
Synonyms: aminoanthraquinone ion(1-) 2-aminoanthraquinone AAQ 2-aminoanthraquinone, ion(1-) ccris 23 nsc 5
Molecular Weight (g/mol): 223.23
Molecular Formula: C14H9NO2
InChi Key: XOGPDSATLSAZEK-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results