accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117-82-8

IUPAC Name: 1,2-bis(2-methoxyethyl) benzene-1,2-dicarboxylate
Synonyms: bis(methoxyethyl)phthalate
Molecular Weight (g/mol): 282.29
Molecular Formula: C14H18O6
InChi Key: HSUIVCLOAAJSRE-UHFFFAOYSA-N
SMILES: COCCOC(=O)C1=CC=CC=C1C(=O)OCCOC
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results