accessibility menu, dialog, popup

UserName

Viewing profile as user:
ICH3NNH3CCH3(E/Z)

CAS RN 117-92-0

IUPAC Name: 2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide
Synonyms: quinaldine red
Molecular Weight (g/mol): 430.33
Molecular Formula: C21H23IN2
InChi Key: JOLANDVPGMEGLK-UHFFFAOYSA-M
SMILES: [I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C
Quantity 
  • (1)
  • (1)
  • (4)
  • (3)
Color 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)