Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Quinaldine red, pure

Catalog No. AC377511000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC377511000 100 g Glass bottle
AC377510050 5 g Glass bottle
AC377510250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC377511000 Supplier Thermo Scientific Chemicals Supplier No. 377511000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 117-92-0 | C21H23IN2 | 430.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-92-0
Molecular Formula C21H23IN2
Molecular Weight (g/mol) 430.33
MDL Number MFCD00011968
InChI Key JOLANDVPGMEGLK-UHFFFAOYSA-M
Synonym 2-[4-(Dimethylamino)styryl]-1-ethylquinolinium iodide
PubChem CID 5702759
SMILES [I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
UV Spectrometry Complies
Color Black/Green or Blue to Gray
Quantity 100 g
Physical Form Powder
IUPAC Name 4-[(E)-2-(1-ethylquinolin-1-ium-2-yl)ethenyl]-N,N-dimethylaniline;iodide
Formula Weight 430.32
Grade Pure
Chemical Name or Material Quinaldine red, Pure

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.