accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1171-47-7

IUPAC Name: 4-[2-(4-carboxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzoic acid
Synonyms: 4-[2-(4-carboxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzoic acid
Molecular Weight (g/mol): 392.25
Molecular Formula: C17H10F6O4
InChi Key: PHQYMDAUTAXXFZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C(C1=CC=C(C=C1)C(O)=O)(C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results