accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SS

CAS RN 1172135-81-7

IUPAC Name: 2-(4-hexylphenyl)-5-[4-(5-phenylthiophen-2-yl)phenyl]thiophene
Synonyms: 2-(4-hexylphenyl)-5-[4-(5-phenylthiophen-2-yl)phenyl]thiophene
Molecular Weight (g/mol): 478.71
Molecular Formula: C32H30S2
InChi Key: SMVIUYXTRPAAAV-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)C1=CC=C(S1)C1=CC=C(C=C1)C1=CC=C(S1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results