Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4″-Hexyl-1,4-bis(5-phenyl-2-thienyl)benzene (purified by sublimation) 99.0+%, TCI America™
SDP

Catalog No. H144850MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H144850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H144850MG Supplier TCI America Supplier No. H144850MG

Chemical Identifiers

CAS 1172135-81-7
Molecular Formula C32H30S2
Molecular Weight (g/mol) 478.71
MDL Number MFCD28100831
InChI Key SMVIUYXTRPAAAV-UHFFFAOYSA-N
Synonym 4′C′C′C-Hexyl-2,2′C-(1,4-phenylene)bis(5-phenylthiophene), 2-(4-Hexylphenyl)-5-[4-(5-phenyl-2-thienyl)phenyl]thiophene, AC5-Hx
PubChem CID 44197204
IUPAC Name 2-(4-hexylphenyl)-5-[4-(5-phenylthiophen-2-yl)phenyl]thiophene
SMILES CCCCCCC1=CC=C(C=C1)C1=CC=C(S1)C1=CC=C(C=C1)C1=CC=C(S1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 50 mg
Formula Weight 478.71
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4′′-Hexyl-1,4-bis(5-phenyl-2-thienyl)benzene (purified by sublimation)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.